The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290325.1 | NP_001277254.1 | 3 - 4 | 1470 | 89 |
NM_001290326.1 | NP_001277255.1 | 3 - 4 | 1470 | 89 | |
NM_032557.6 | NP_115946.2 | 3 - 4 | 1470 | 89 | |
GenBank mRNA | AB067478.1 | - | 2 - 3 | 151 | 89 |
AK093754.1 | - | 3 - 4 | 693 | 89 | |
AK126943.1 | - | 4 - 5 | 1298 | 89 | |
AL833976.1 | - | 3 - 4 | 1442 | 89 | |
BC039115.1 | - | 3 - 4 | 1470 | 89 | |
BC068975.1 | - | 3 - 4 | 1417 | 89 |