The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290325.1 | NP_001277254.1 | 6 - 7 | 1935 | 168 |
NM_001290326.1 | NP_001277255.1 | 7 - 8 | 2051 | 168 | |
NM_032557.6 | NP_115946.2 | 6 - 7 | 1935 | 168 | |
XM_011532360.2 | XP_011530662.1 | 5 - 6 | 1321 | 168 | |
GenBank mRNA | AB067478.1 | - | 5 - 6 | 616 | 168 |
AK057992.1 | - | 3 - 4 | 459 | 168 | |
AK093754.1 | - | 7 - 8 | 1274 | 168 | |
AK126943.1 | - | 7 - 8 | 1763 | 168 | |
AL833976.1 | - | 6 - 7 | 1907 | 168 | |
BC039115.1 | - | 6 - 7 | 1935 | 168 | |
BC068975.1 | - | 6 - 7 | 1882 | 168 |