Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MORN5
Entrez Gene ID
254956
Gene Name
MORN repeat containing 5
Gene Aliases
C9orf113, C9orf18
Chromosome Location
Chr.9: 122159661 - 122200088 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005251878.3 XP_005251935.1 1 - 2 84 72

View Full Product Details