Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TNRC6B
Entrez Gene ID
23112
Gene Name
trinucleotide repeat containing 6B
Gene Aliases
-
Chromosome Location
Chr.22: 40044817 - 40335808 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024843.1 NP_001020014.1 11 - 12 1323 82
NM_001162501.1 NP_001155973.1 9 - 10 3473 82
NM_015088.2 NP_055903.2 8 - 9 3314 82
GenBank mRNA AB029016.2 - 8 - 9 3297 82
AK294519.1 - 9 - 10 3464 82
AK304087.1 - 5 - 6 799 82
BC028626.2 - 11 - 12 1256 82

View Full Product Details