Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TNRC6B
Entrez Gene ID
23112
Gene Name
trinucleotide repeat containing 6B
Gene Aliases
-
Chromosome Location
Chr.22: 40044817 - 40335808 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024843.1 NP_001020014.1 14 - 15 1724 60
NM_001162501.1 NP_001155973.1 13 - 14 4045 60
NM_015088.2 NP_055903.2 11 - 12 3715 60
GenBank mRNA AB029016.2 - 11 - 12 3698 60
AK294519.1 - 13 - 14 4036 60
AK304087.1 - 8 - 9 1200 60
BC028626.2 - 14 - 15 1657 60
BX647630.1 - 8 - 9 2917 60

View Full Product Details