The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001169122.1 | NP_001162593.1 | 9 - 10 | 1936 | 81 |
NM_001169123.1 | NP_001162594.1 | 10 - 11 | 2005 | 81 | |
NM_001169124.1 | NP_001162595.1 | 9 - 10 | 1930 | 81 | |
NM_001169125.1 | NP_001162596.1 | 9 - 10 | 1918 | 81 | |
NM_001170628.1 | NP_001164099.1 | 7 - 8 | 569 | 81 | |
NM_002025.3 | NP_002016.2 | 10 - 11 | 2035 | 81 | |
GenBank mRNA | AB102644.1 | - | 9 - 10 | 1439 | 81 |
AJ001550.1 | - | 9 - 10 | 1439 | 81 | |
AK301927.1 | - | 7 - 8 | 569 | 81 | |
BC132683.1 | - | 10 - 11 | 1577 | 81 | |
BC143740.1 | - | 9 - 10 | 1490 | 81 | |
BC143744.1 | - | 9 - 10 | 1502 | 81 | |
L76569.1 | - | 9 - 10 | 1849 | 81 | |
U48436.1 | - | 10 - 11 | 2035 | 81 |