Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AURKB
Entrez Gene ID
9212
Gene Name
aurora kinase B
Gene Aliases
AIK2, AIM-1, AIM1, ARK2, AurB, IPL1, PPP1R48, STK12, STK5, aurkb-sv1, aurkb-sv2
Chromosome Location
Chr.17: 8204731 - 8210768 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001284526.1 NP_001271455.1 4 - 5 331 84
NM_001313952.1 NP_001300881.1 3 - 4 319 84
NR_132731.1 3 - 4 210 84
GenBank mRNA AB519679.1 - 3 - 4 209 84
AY677083.1 - 3 - 4 210 84
BC013300.2 - 4 - 5 287 84
BM905813.1 - 3 - 4 280 84

View Full Product Details