The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256834.2 | NP_001243763.1 | 7 - 8 | 916 | 109 |
NM_001284526.1 | NP_001271455.1 | 8 - 9 | 992 | 109 | |
NM_001313950.1 | NP_001300879.1 | 8 - 9 | 965 | 109 | |
NM_001313951.1 | NP_001300880.1 | 8 - 9 | 994 | 109 | |
NM_001313952.1 | NP_001300881.1 | 7 - 8 | 980 | 109 | |
NM_001313953.1 | NP_001300882.1 | 7 - 8 | 772 | 109 | |
NM_001313954.1 | NP_001300883.1 | 8 - 9 | 999 | 109 | |
NM_001313955.1 | NP_001300884.1 | 6 - 7 | 635 | 109 | |
NM_004217.3 | NP_004208.2 | 8 - 9 | 989 | 109 | |
NR_132730.1 | 8 - 9 | 847 | 109 | ||
NR_132731.1 | 6 - 7 | 732 | 109 | ||
XM_011524070.2 | XP_011522372.1 | 8 - 9 | 842 | 109 | |
XM_011524072.2 | XP_011522374.1 | 7 - 8 | 1105 | 109 | |
XM_017025307.1 | XP_016880796.1 | 7 - 8 | 1032 | 109 | |
XM_017025308.1 | XP_016880797.1 | 7 - 8 | 962 | 109 | |
XM_017025309.1 | XP_016880798.1 | 7 - 8 | 870 | 109 | |
XM_017025310.1 | XP_016880799.1 | 8 - 9 | 852 | 109 | |
XM_017025311.1 | XP_016880800.1 | 4 - 5 | 529 | 109 | |
GenBank mRNA | AB011446.1 | - | 8 - 9 | 930 | 109 |
AB011450.1 | - | 8 - 9 | 924 | 109 | |
AB519677.1 | - | 7 - 8 | 867 | 109 | |
AB519678.1 | - | 7 - 8 | 771 | 109 | |
AB519679.1 | - | 6 - 7 | 731 | 109 | |
AF004022.1 | - | 8 - 9 | 934 | 109 | |
AF008552.1 | - | 7 - 8 | 867 | 109 | |
AF015254.1 | - | 8 - 9 | 934 | 109 | |
AK297976.1 | - | 7 - 8 | 871 | 109 | |
AK314889.1 | - | 8 - 9 | 938 | 109 | |
AY677083.1 | - | 6 - 7 | 732 | 109 | |
BC000442.2 | - | 8 - 9 | 928 | 109 | |
BC009751.2 | - | 8 - 9 | 908 | 109 | |
BC013300.2 | - | 8 - 9 | 948 | 109 | |
BC080581.1 | - | 8 - 9 | 920 | 109 | |
BT019534.1 | - | 7 - 8 | 867 | 109 |