The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000796.5 | NP_000787.2 | 2 - 3 | 703 | 63 |
NM_001282563.2 | NP_001269492.1 | 3 - 4 | 562 | 63 | |
NM_001290809.1 | NP_001277738.1 | 3 - 4 | 695 | 63 | |
NM_033663.5 | NP_387512.3 | 2 - 3 | 703 | 63 | |
XM_017005829.1 | XP_016861318.1 | 3 - 4 | 523 | 63 | |
GenBank mRNA | AK292309.1 | - | 3 - 4 | 562 | 63 |
BC095510.1 | - | 1 - 2 | 273 | 63 | |
BC128123.1 | - | 2 - 3 | 349 | 63 | |
KJ534829.1 | - | 1 - 2 | 272 | 63 | |
L20469.1 | - | 1 - 2 | 272 | 63 | |
U32499.1 | - | 1 - 2 | 272 | 63 |