Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- LINC00852
- Entrez Gene ID
-
84657
- Gene Name
- long intergenic non-protein coding RNA 852
- Gene Aliases
-
C3orf42, GHRL-AS2, GHRLOS2, NAG73
- Chromosome Location
-
Chr.3: 10284419 - 10285746 on Build GRCh38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BG491277.1 |
- |
1 |
508 |
|
Chr. 3 - 10284467 - 10285107 [+] on Build GRCh38 |
100 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
ghrelin opposite strand/antisense RNA |
GHRLOS |
GHRL-AS1, GHRLAS, NCRNA00068 |
|
Sense |
Chr. - 10280952 - 10293449 on Build GRCh38 |
Downstream |
ghrelin/obestatin prepropeptide |
GHRL |
MTLRP |
643.0
|
Anti-sense |
Chr. - 10285750 - 10292947 on Build GRCh38 |
View Full Product Details