The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080497.2 | NP_001073966.2 | 4 - 5 | 1210 | 70 |
GenBank mRNA | AB011542.1 | - | 3 - 4 | 402 | 70 |
AK295380.1 | - | 4 - 5 | 1210 | 70 |