Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RBM18
Entrez Gene ID
92400
Gene Name
RNA binding motif protein 18
Gene Aliases
-
Chromosome Location
Chr.9: 122239555 - 122264864 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033117.3 NP_149108.1 1 - 2 149 70
NR_027125.1 1 - 2 149 70
NR_027126.1 1 - 2 149 70
GenBank mRNA AK057676.1 - 1 - 2 125 70
AK301050.1 - 1 - 2 105 70
AK310008.1 - 1 - 2 106 70
AK310807.1 - 1 - 2 91 70
AL389986.1 - 1 - 2 77 70
BC008942.2 - 1 - 2 96 70
BC019319.1 - 1 - 2 96 70
BP870502.1 - 1 - 2 128 70
DA308339.1 - 1 - 2 149 70

View Full Product Details