The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001197238.1 | NP_001184167.1 | 1 - 2 | 202 | 112 |
XM_005248797.3 | XP_005248854.1 | 1 - 2 | 96 | 112 | |
XM_005248798.4 | XP_005248855.1 | 1 - 2 | 404 | 112 | |
XM_006714955.3 | XP_006715018.1 | 1 - 2 | 96 | 112 | |
XM_011514229.2 | XP_011512531.1 | 1 - 2 | 96 | 112 | |
XM_011514231.2 | XP_011512533.1 | 1 - 2 | 404 | 112 | |
GenBank mRNA | BC017497.1 | - | 1 - 2 | 94 | 112 |
BP234194.1 | - | 1 - 2 | 202 | 112 |