The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042549.1 | NP_001036014.1 | 1 - 2 | 264 | 110 |
NM_001297736.1 | NP_001284665.1 | 1 - 2 | 264 | 110 | |
NM_001297737.1 | NP_001284666.1 | 1 - 2 | 264 | 110 | |
NM_001297739.1 | NP_001284668.1 | 1 - 2 | 264 | 110 | |
NM_015471.3 | NP_056286.3 | 1 - 2 | 264 | 110 | |
GenBank mRNA | AF201941.1 | - | 1 - 2 | 236 | 110 |
AF255793.1 | - | 1 - 2 | 250 | 110 | |
AI703127.1 | - | 1 - 2 | 48 | 110 | |
AK222996.1 | - | 1 - 2 | 234 | 110 | |
AK223260.1 | - | 1 - 2 | 244 | 110 | |
AK303250.1 | - | 1 - 2 | 244 | 110 | |
AK313730.1 | - | 1 - 2 | 260 | 110 | |
AL050084.1 | - | 1 - 2 | 43 | 110 | |
BC007067.1 | - | 1 - 2 | 238 | 110 | |
BC036048.1 | - | 1 - 2 | 242 | 110 | |
BE669670.1 | - | 1 - 2 | 69 | 110 | |
BU570983.1 | - | 1 - 2 | 244 | 110 | |
CK003923.1 | - | 1 - 2 | 247 | 110 | |
DB045932.1 | - | 1 - 2 | 264 | 110 |