The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163334.1 | NP_001156806.1 | 14 - 15 | 2127 | 81 |
NM_001163335.1 | NP_001156807.1 | 15 - 16 | 2239 | 81 | |
NM_138780.2 | NP_620135.1 | 14 - 15 | 2148 | 81 | |
XM_011544001.2 | XP_011542303.1 | 17 - 18 | 2566 | 81 | |
XM_011544002.2 | XP_011542304.1 | 15 - 16 | 2200 | 81 | |
XM_017029972.1 | XP_016885461.1 | 18 - 19 | 2732 | 81 | |
XM_017029973.1 | XP_016885462.1 | 17 - 18 | 2799 | 81 | |
XM_017029974.1 | XP_016885463.1 | 16 - 17 | 2427 | 81 | |
GenBank mRNA | AB080222.1 | - | 13 - 14 | 1705 | 81 |
AK125106.1 | - | 14 - 15 | 2116 | 81 | |
BC131585.1 | - | 14 - 15 | 1771 | 81 | |
BX647688.1 | - | 15 - 16 | 2241 | 81 |