Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYTL5
Entrez Gene ID
94122
Gene Name
synaptotagmin like 5
Gene Aliases
slp5
Chromosome Location
Chr.X: 37888915 - 38128820 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163335.1 NP_001156807.1 11 - 12 1694 74
NM_138780.2 NP_620135.1 10 - 11 1603 74
GenBank mRNA AB080222.1 - 9 - 10 1160 74
AK125106.1 - 10 - 11 1571 74
BX647688.1 - 11 - 12 1696 74

View Full Product Details