The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001307940.1 | NP_001294869.1 | 15 - 16 | 1799 | 109 |
NM_024921.3 | NP_079197.3 | 15 - 16 | 1799 | 109 | |
XM_005262203.3 | XP_005262260.1 | 15 - 16 | 1712 | 109 | |
GenBank mRNA | AF309774.1 | - | 15 - 16 | 1799 | 109 |
AK025080.1 | - | 7 - 8 | 764 | 109 | |
AK026445.1 | - | 3 - 4 | 292 | 109 | |
AK225193.1 | - | 7 - 8 | 764 | 109 | |
AK290360.1 | - | 15 - 16 | 1748 | 109 | |
AK314295.1 | - | 15 - 16 | 1718 | 109 | |
BC017500.1 | - | 15 - 16 | 1773 | 109 |