The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001307940.1 | NP_001294869.1 | 6 - 7 | 865 | 108 |
NM_024921.3 | NP_079197.3 | 6 - 7 | 865 | 108 | |
XM_005262203.3 | XP_005262260.1 | 6 - 7 | 778 | 108 | |
GenBank mRNA | AF309774.1 | - | 6 - 7 | 865 | 108 |
AK025039.1 | - | 6 - 7 | 814 | 108 | |
AK290360.1 | - | 6 - 7 | 814 | 108 | |
AK314295.1 | - | 6 - 7 | 784 | 108 | |
BC017500.1 | - | 6 - 7 | 839 | 108 |