The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007240.2 | NP_001007241.2 | 10 - 11 | 1700 | 107 |
NM_001007241.2 | NP_001007242.2 | 9 - 10 | 1259 | 107 | |
NM_001007242.2 | NP_001007243.2 | 8 - 9 | 1250 | 107 | |
NM_001502.3 | NP_001493.2 | 9 - 10 | 1691 | 107 | |
XM_017023155.1 | XP_016878644.1 | 9 - 10 | 1444 | 107 | |
GenBank mRNA | AB035541.1 | - | 8 - 9 | 1480 | 107 |
AB035542.1 | - | 7 - 8 | 1039 | 107 | |
AK293384.1 | - | 9 - 10 | 1447 | 107 | |
AK312277.1 | - | 9 - 10 | 1578 | 107 | |
BC032693.2 | - | 7 - 8 | 1097 | 107 | |
BX647831.1 | - | 10 - 11 | 1588 | 107 | |
BX648023.1 | - | 9 - 10 | 1585 | 107 | |
BX648201.1 | - | 8 - 9 | 1137 | 107 | |
BX649067.1 | - | 9 - 10 | 1146 | 107 | |
CR749600.1 | - | 9 - 10 | 1650 | 107 | |
U36221.1 | - | 9 - 10 | 1560 | 107 |