The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_201567.1 | NP_963861.1 | 5 - 6 | 797 | 74 |
XM_006713435.2 | XP_006713498.1 | 5 - 6 | 889 | 74 | |
GenBank mRNA | AF277722.1 | - | 5 - 6 | 325 | 74 |
AY137580.1 | - | 5 - 6 | 433 | 74 | |
EU812118.1 | - | 5 - 6 | 430 | 74 |