The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003846.2 | NP_003837.1 | 1 - 2 | 298 | 76 |
GenBank mRNA | AB018080.1 | - | 1 - 2 | 80 | 76 |
AF093670.1 | - | 1 - 2 | 78 | 76 | |
AK023991.1 | - | 1 - 2 | 62 | 76 | |
BC011963.1 | - | 1 - 2 | 78 | 76 | |
CR542047.1 | - | 1 - 2 | 62 | 76 | |
DB054933.1 | - | 1 - 2 | 298 | 76 |