The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133492.2 | NP_597999.1 | 2 - 3 | 287 | 69 |
XM_011527673.2 | XP_011525975.1 | 3 - 4 | 425 | 69 | |
GenBank mRNA | AF347024.1 | - | 2 - 3 | 209 | 69 |
BC112122.1 | - | 2 - 3 | 287 | 69 | |
BC112124.1 | - | 2 - 3 | 287 | 69 |