Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- NEB
- Entrez Gene ID
-
4703
- Gene Name
- nebulin
- Gene Aliases
-
NEB177D, NEM2
- Chromosome Location
-
Chr.2: 151485334 - 151734487 on Build GRCh38
View Full Product Details