Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NEB
Entrez Gene ID
4703
Gene Name
nebulin
Gene Aliases
NEB177D, NEM2
Chromosome Location
Chr.2: 151485334 - 151734487 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164507.1 NP_001157979.1 72 - 73 10762 94
NM_001164508.1 NP_001157980.1 72 - 73 10762 94
NM_001271208.1 NP_001258137.1 72 - 73 10762 94
NM_004543.4 NP_004534.2 69 - 70 10033 94
XM_005246590.2 XP_005246647.1 72 - 73 10749 94
XM_005246591.2 XP_005246648.1 72 - 73 10749 94
XM_005246592.2 XP_005246649.1 72 - 73 10749 94
XM_005246593.2 XP_005246650.1 72 - 73 10749 94
XM_005246594.2 XP_005246651.1 72 - 73 10749 94
XM_005246596.2 XP_005246653.1 72 - 73 10749 94
XM_005246597.2 XP_005246654.1 72 - 73 10749 94
XM_005246598.2 XP_005246655.1 72 - 73 10749 94
XM_005246599.2 XP_005246656.1 72 - 73 10749 94
XM_005246600.2 XP_005246657.1 72 - 73 10749 94
XM_005246601.2 XP_005246658.1 72 - 73 10749 94
XM_005246602.2 XP_005246659.1 72 - 73 10749 94
XM_005246603.2 XP_005246660.1 72 - 73 10749 94
XM_005246604.2 XP_005246661.1 72 - 73 10749 94
XM_005246606.2 XP_005246663.1 72 - 73 10749 94
XM_005246608.2 XP_005246665.1 72 - 73 10749 94
XM_005246610.2 XP_005246667.1 72 - 73 10749 94
XM_005246611.2 XP_005246668.1 72 - 73 10749 94
XM_005246612.2 XP_005246669.1 68 - 69 10020 94
XM_005246613.2 XP_005246670.1 69 - 70 10020 94
XM_005246615.2 XP_005246672.1 72 - 73 10749 94
XM_005246616.1 XP_005246673.1 72 - 73 10749 94
XM_006712541.2 XP_006712604.1 72 - 73 10749 94
XM_006712542.2 XP_006712605.1 72 - 73 10749 94
XM_011511225.2 XP_011509527.1 72 - 73 10749 94
XM_011511226.2 XP_011509528.1 61 - 62 8562 94
XM_017004177.1 XP_016859666.1 72 - 73 10749 94
XM_017004178.1 XP_016859667.1 72 - 73 10749 94
XM_017004179.1 XP_016859668.1 72 - 73 10749 94
XM_017004180.1 XP_016859669.1 72 - 73 10749 94
XM_017004181.1 XP_016859670.1 72 - 73 10749 94
XM_017004182.1 XP_016859671.1 72 - 73 10749 94
XM_017004183.1 XP_016859672.1 72 - 73 10749 94
XM_017004184.1 XP_016859673.1 72 - 73 10749 94
XM_017004185.1 XP_016859674.1 72 - 73 10749 94
GenBank mRNA BP233123.1 - 2 - 3 219 94

View Full Product Details