Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NEB
Entrez Gene ID
4703
Gene Name
nebulin
Gene Aliases
NEB177D, NEM2
Chromosome Location
Chr.2: 151485334 - 151734487 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164507.1 NP_001157979.1 110 - 111 17732 87
NM_001164508.1 NP_001157980.1 110 - 111 17732 87
NM_001271208.1 NP_001258137.1 110 - 111 17732 87
NM_004543.4 NP_004534.2 83 - 84 12629 87
XM_005246590.2 XP_005246647.1 110 - 111 17719 87
XM_005246591.2 XP_005246648.1 110 - 111 17719 87
XM_005246592.2 XP_005246649.1 110 - 111 17719 87
XM_005246593.2 XP_005246650.1 110 - 111 17719 87
XM_005246594.2 XP_005246651.1 110 - 111 17719 87
XM_005246596.2 XP_005246653.1 110 - 111 17719 87
XM_005246597.2 XP_005246654.1 110 - 111 17719 87
XM_005246598.2 XP_005246655.1 110 - 111 17719 87
XM_005246599.2 XP_005246656.1 110 - 111 17719 87
XM_005246600.2 XP_005246657.1 110 - 111 17719 87
XM_005246601.2 XP_005246658.1 110 - 111 17719 87
XM_005246602.2 XP_005246659.1 110 - 111 17719 87
XM_005246603.2 XP_005246660.1 110 - 111 17719 87
XM_005246604.2 XP_005246661.1 110 - 111 17719 87
XM_005246606.2 XP_005246663.1 110 - 111 17719 87
XM_005246608.2 XP_005246665.1 110 - 111 17719 87
XM_005246610.2 XP_005246667.1 110 - 111 17719 87
XM_005246611.2 XP_005246668.1 110 - 111 17719 87
XM_005246612.2 XP_005246669.1 106 - 107 16990 87
XM_005246613.2 XP_005246670.1 107 - 108 16990 87
XM_005246615.2 XP_005246672.1 110 - 111 17719 87
XM_005246616.1 XP_005246673.1 110 - 111 17719 87
XM_005246617.2 XP_005246674.1 94 - 95 14803 87
XM_006712541.2 XP_006712604.1 110 - 111 17719 87
XM_006712542.2 XP_006712605.1 110 - 111 17719 87
XM_011511225.2 XP_011509527.1 110 - 111 17719 87
XM_011511226.2 XP_011509528.1 99 - 100 15532 87
XM_011511227.2 XP_011509529.1 86 - 87 13345 87
XM_017004177.1 XP_016859666.1 110 - 111 17719 87
XM_017004178.1 XP_016859667.1 110 - 111 17719 87
XM_017004179.1 XP_016859668.1 110 - 111 17719 87
XM_017004180.1 XP_016859669.1 110 - 111 17719 87
XM_017004181.1 XP_016859670.1 110 - 111 17719 87
XM_017004182.1 XP_016859671.1 110 - 111 17719 87
XM_017004183.1 XP_016859672.1 110 - 111 17719 87
XM_017004184.1 XP_016859673.1 110 - 111 17719 87
XM_017004185.1 XP_016859674.1 110 - 111 17719 87
GenBank mRNA X83957.1 - 70 - 71 12866 87

View Full Product Details