The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130438.2 | NP_001123910.1 | 17 - 18 | 2578 | 84 |
NM_001195532.1 | NP_001182461.1 | 17 - 18 | 2578 | 84 | |
NM_003127.3 | NP_003118.2 | 17 - 18 | 2578 | 84 | |
XM_006717245.1 | XP_006717308.1 | 17 - 18 | 2485 | 84 | |
XM_006717246.1 | XP_006717309.1 | 17 - 18 | 2485 | 84 | |
XM_006717247.2 | XP_006717310.1 | 17 - 18 | 2485 | 84 | |
XM_006717248.2 | XP_006717311.1 | 17 - 18 | 2484 | 84 | |
XM_006717249.1 | XP_006717312.1 | 17 - 18 | 2485 | 84 | |
XM_006717250.1 | XP_006717313.1 | 17 - 18 | 2485 | 84 | |
XM_006717251.2 | XP_006717314.1 | 17 - 18 | 2572 | 84 | |
XM_006717252.2 | XP_006717315.1 | 17 - 18 | 2485 | 84 | |
XM_006717253.1 | XP_006717316.1 | 17 - 18 | 2485 | 84 | |
XM_006717254.2 | XP_006717317.1 | 17 - 18 | 2572 | 84 | |
XM_017015059.1 | XP_016870548.1 | 17 - 18 | 2563 | 84 | |
XM_017015060.1 | XP_016870549.1 | 17 - 18 | 2485 | 84 | |
XM_017015061.1 | XP_016870550.1 | 17 - 18 | 2543 | 84 | |
GenBank mRNA | AB191262.1 | - | 17 - 18 | 2543 | 84 |
AB209860.1 | - | 17 - 18 | 2540 | 84 | |
AK294753.1 | - | 18 - 19 | 2602 | 84 | |
AK300386.1 | - | 17 - 18 | 2546 | 84 | |
AK307676.1 | - | 17 - 18 | 2546 | 84 | |
BC053521.1 | - | 17 - 18 | 2527 | 84 | |
J05243.1 | - | 17 - 18 | 2538 | 84 | |
M18627.1 | - | 2 - 3 | 412 | 84 | |
M24773.1 | - | 2 - 3 | 412 | 84 | |
U83867.1 | - | 17 - 18 | 2505 | 84 |