Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPTAN1
Entrez Gene ID
6709
Gene Name
spectrin alpha, non-erythrocytic 1
Gene Aliases
EIEE5, NEAS, SPTA2
Chromosome Location
Chr.9: 128552558 - 128633665 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130438.2 NP_001123910.1 56 - 57 7451 69
NM_001195532.1 NP_001182461.1 54 - 55 7376 69
NM_003127.3 NP_003118.2 55 - 56 7436 69
XM_006717245.1 XP_006717308.1 57 - 58 7421 69
XM_006717246.1 XP_006717309.1 56 - 57 7406 69
XM_006717247.2 XP_006717310.1 56 - 57 7361 69
XM_006717248.2 XP_006717311.1 56 - 57 7357 69
XM_006717249.1 XP_006717312.1 55 - 56 7343 69
XM_006717250.1 XP_006717313.1 55 - 56 7340 69
XM_006717251.2 XP_006717314.1 56 - 57 7448 69
XM_006717252.2 XP_006717315.1 55 - 56 7298 69
XM_006717253.1 XP_006717316.1 54 - 55 7283 69
XM_006717254.2 XP_006717317.1 57 - 58 7508 69
XM_017015059.1 XP_016870548.1 55 - 56 7418 69
XM_017015060.1 XP_016870549.1 54 - 55 7280 69
XM_017015061.1 XP_016870550.1 55 - 56 7356 69
GenBank mRNA AB191262.1 - 54 - 55 7341 69
AB209860.1 - 55 - 56 7398 69
AK294753.1 - 32 - 33 3980 69
AL110273.1 - 34 - 35 4263 69
BC053521.1 - 54 - 55 7325 69
J05243.1 - 55 - 56 7396 69
U83867.1 - 56 - 57 7378 69

View Full Product Details