The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170538.1 | NP_001164009.1 | 7 - 8 | 1307 | 63 |
NM_173543.2 | NP_775814.2 | 6 - 7 | 1360 | 63 | |
XM_005247198.3 | XP_005247255.1 | 6 - 7 | 2470 | 63 | |
XM_005247200.4 | XP_005247257.1 | 2 - 3 | 1395 | 63 | |
XM_006713526.1 | XP_006713589.1 | 7 - 8 | 1706 | 63 | |
XM_006713527.3 | XP_006713590.1 | 6 - 7 | 1613 | 63 | |
XM_011512518.2 | XP_011510820.1 | 6 - 7 | 2470 | 63 | |
XM_011512520.2 | XP_011510822.1 | 7 - 8 | 1617 | 63 | |
XM_011512521.2 | XP_011510823.1 | 7 - 8 | 2767 | 63 | |
XM_011512522.1 | XP_011510824.1 | 7 - 8 | 1307 | 63 | |
XM_011512524.2 | XP_011510826.1 | 6 - 7 | 2470 | 63 | |
XM_011512528.2 | XP_011510830.1 | 6 - 7 | 2470 | 63 | |
XM_017005841.1 | XP_016861330.1 | 6 - 7 | 2470 | 63 | |
XM_017005842.1 | XP_016861331.1 | 6 - 7 | 2470 | 63 | |
GenBank mRNA | AK057406.1 | - | 7 - 8 | 1307 | 63 |
BC033308.1 | - | 6 - 7 | 1360 | 63 |