The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017577.4 | NP_060047.3 | 4 - 5 | 435 | 155 |
XM_005247547.1 | XP_005247604.1 | 4 - 5 | 397 | 155 | |
XM_011512930.1 | XP_011511232.1 | 4 - 5 | 430 | 155 | |
XM_017006647.1 | XP_016862136.1 | 4 - 5 | 425 | 155 | |
GenBank mRNA | AK097783.1 | - | 4 - 5 | 431 | 155 |
AK292846.1 | - | 4 - 5 | 371 | 155 | |
BC035040.1 | - | 4 - 5 | 441 | 155 |