The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016243.2 | NP_057327.2 | 4 - 5 | 419 | 135 |
GenBank mRNA | AF087912.1 | - | 4 - 5 | 386 | 135 |
AF093822.1 | - | 4 - 5 | 386 | 135 | |
AF125533.1 | - | 4 - 5 | 386 | 135 | |
AF169481.1 | - | 4 - 5 | 374 | 135 | |
AK074654.1 | - | 4 - 5 | 400 | 135 | |
AK075304.1 | - | 4 - 5 | 368 | 135 | |
AK223416.1 | - | 4 - 5 | 400 | 135 | |
AK295902.1 | - | 4 - 5 | 552 | 135 | |
AK313333.1 | - | 4 - 5 | 398 | 135 | |
AY359026.1 | - | 4 - 5 | 423 | 135 | |
BC018732.1 | - | 4 - 5 | 376 | 135 | |
BC127945.1 | - | 1 - 2 | 106 | 135 |