The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001165412.1 | NP_001158884.1 | 21 - 22 | 2886 | 89 |
NM_001319226.1 | NP_001306155.1 | 21 - 22 | 2696 | 89 | |
NM_003998.3 | NP_003989.2 | 21 - 22 | 2889 | 89 | |
XM_011532006.2 | XP_011530308.1 | 22 - 23 | 2656 | 89 | |
XM_011532008.2 | XP_011530310.1 | 19 - 20 | 2270 | 89 | |
GenBank mRNA | AK122850.1 | - | 14 - 15 | 3018 | 89 |
AK291450.1 | - | 21 - 22 | 2550 | 89 | |
BC033210.1 | - | 9 - 10 | 1167 | 89 | |
BC051765.1 | - | 21 - 22 | 2889 | 89 | |
CR749522.1 | - | 21 - 22 | 2646 | 89 | |
M55643.1 | - | 21 - 22 | 2603 | 89 | |
M58603.1 | - | 21 - 22 | 2816 | 89 |