The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018727.5 | NP_061197.4 | 10 - 11 | 1705 | 59 |
NM_080704.3 | NP_542435.2 | 11 - 12 | 1822 | 59 | |
NM_080705.3 | NP_542436.2 | 10 - 11 | 1748 | 59 | |
NM_080706.3 | NP_542437.2 | 9 - 10 | 2074 | 59 | |
GenBank mRNA | AF196175.1 | - | 10 - 11 | 1760 | 59 |
AF196176.1 | - | 10 - 11 | 1746 | 59 | |
AJ272063.2 | - | 10 - 11 | 1705 | 59 | |
AJ277028.1 | - | 9 - 10 | 2074 | 59 | |
AL136801.1 | - | 11 - 12 | 1818 | 59 | |
AY131289.1 | - | 9 - 10 | 1547 | 59 | |
AY986821.1 | - | 8 - 9 | 1367 | 59 | |
BC132820.1 | - | 9 - 10 | 2026 | 59 | |
BC136633.1 | - | 9 - 10 | 2026 | 59 | |
DQ177332.1 | - | 8 - 9 | 1634 | 59 | |
DQ177333.1 | - | 10 - 11 | 1597 | 59 |