The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001083899.2 | NP_001077368.2 | 6 - 7 | 750 | 93 |
NM_001256017.2 | NP_001242946.2 | 5 - 6 | 696 | 93 | |
NM_016363.5 | NP_057447.5 | 6 - 7 | 750 | 93 | |
GenBank mRNA | AB035073.1 | - | 6 - 7 | 734 | 93 |
AB043819.1 | - | 6 - 7 | 750 | 93 | |
AB043820.1 | - | 5 - 6 | 696 | 93 | |
AB043821.1 | - | 6 - 7 | 750 | 93 | |
BC104832.1 | - | 6 - 7 | 742 | 93 | |
BC111963.1 | - | 6 - 7 | 742 | 93 | |
BQ216894.1 | - | 1 - 2 | 177 | 93 |