The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173481.3 | NP_775752.1 | 1 - 2 | 61 | 81 |
GenBank mRNA | BC052236.1 | - | 1 - 2 | 48 | 81 |