The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021073.2 | NP_066551.1 | 1 - 2 | 1208 | 82 |
XM_005249304.3 | XP_005249361.1 | 1 - 2 | 1256 | 82 | |
XM_011514816.2 | XP_011513118.1 | 1 - 2 | 504 | 82 | |
XM_011514817.2 | XP_011513119.1 | 1 - 2 | 1256 | 82 | |
GenBank mRNA | AK291559.1 | - | 1 - 2 | 1221 | 82 |
AK303576.1 | - | 1 - 2 | 510 | 82 | |
BC027958.1 | - | 1 - 2 | 1208 | 82 | |
KC485577.1 | - | 1 - 2 | 496 | 82 | |
M60314.1 | - | 1 - 2 | 1194 | 82 |