The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018310.3 | NP_060780.2 | 2 - 3 | 344 | 97 |
GenBank mRNA | AF206673.2 | - | 2 - 3 | 344 | 97 |
AF298153.1 | - | 2 - 3 | 267 | 97 | |
AK001914.1 | - | 2 - 3 | 306 | 97 | |
AK294337.1 | - | 3 - 4 | 303 | 97 | |
AK315420.1 | - | 2 - 3 | 293 | 97 | |
BC010648.1 | - | 2 - 3 | 297 | 97 | |
DA688249.1 | - | 2 - 3 | 344 | 97 |