The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015881.5 | NP_056965.3 | 1 - 2 | 214 | 61 |
XM_017017554.1 | XP_016873043.1 | 1 - 2 | 179 | 61 | |
GenBank mRNA | BC007660.2 | - | 1 - 2 | 213 | 61 |
CB156451.1 | - | 1 - 2 | 187 | 61 |