Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SFTPC
Entrez Gene ID
6440
Gene Name
surfactant protein C
Gene Aliases
BRICD6, PSP-C, SFTP2, SMDP2, SP-C
Chromosome Location
Chr.8: 22161671 - 22164479 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172410.1 NP_001165881.1 4 - 5 594 105
NM_001317779.1 NP_001304708.1 3 - 4 435 105
NM_003018.3 NP_003009.2 4 - 5 594 105
XM_011544613.2 XP_011542915.1 3 - 4 374 105
GenBank mRNA AK315742.1 - 4 - 5 468 105
BC005913.1 - 4 - 5 467 105
BI490051.1 - 3 - 4 299 105
BM993053.1 - 2 - 3 105
BQ027183.1 - 2 - 3 105
J03517.1 - 4 - 5 463 105
J03553.1 - 4 - 5 610 105
KU178329.1 - 4 - 5 436 105

View Full Product Details