The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006417.4 | NP_006408.3 | 8 - 9 | 81 | |
NR_135641.1 | 7 - 8 | 81 | |||
GenBank mRNA | BC022870.1 | - | 8 - 9 | 81 | |
CA426666.1 | - | 1 - 2 | 328 | 81 |