Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRKCZ
Entrez Gene ID
5590
Gene Name
protein kinase C zeta
Gene Aliases
PKC-ZETA, PKC2
Chromosome Location
Chr.1: 2050470 - 2185395 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033581.1 NP_001028753.1 3 - 4 499 88
NM_001033582.1 NP_001028754.1 3 - 4 396 88
NM_001242874.1 NP_001229803.1 3 - 4 506 88
NM_002744.4 NP_002735.3 6 - 7 711 88
XM_011541773.1 XP_011540075.1 6 - 7 707 88
XM_011541774.1 XP_011540076.1 3 - 4 572 88
XM_011541775.2 XP_011540077.1 6 - 7 707 88
XM_011541776.1 XP_011540078.1 3 - 4 512 88
XM_011541778.1 XP_011540080.1 3 - 4 425 88
XM_017001789.1 XP_016857278.1 6 - 7 707 88
XM_017001790.1 XP_016857279.1 6 - 7 707 88
XM_017001791.1 XP_016857280.1 3 - 4 356 88
XM_017001792.1 XP_016857281.1 3 - 4 512 88
XM_017001793.1 XP_016857282.1 3 - 4 770 88
XM_017001794.1 XP_016857283.1 3 - 4 425 88
XM_017001795.1 XP_016857284.1 3 - 4 429 88
XM_017001796.1 XP_016857285.1 3 - 4 512 88
XM_017001798.1 XP_016857287.1 3 - 4 425 88
XM_017001799.1 XP_016857288.1 3 - 4 833 88
XM_017001802.1 XP_016857291.1 3 - 4 512 88
XM_017001803.1 XP_016857292.1 6 - 7 707 88
XM_017001804.1 XP_016857293.1 3 - 4 422 88
XM_017001805.1 XP_016857294.1 3 - 4 392 88
GenBank mRNA AK290995.1 - 6 - 7 550 88
AK294649.1 - 3 - 4 501 88
AK294782.1 - 3 - 4 419 88
AL542576.3 - 3 - 4 386 88
AU134793.2 - 3 - 4 396 88
BC008058.2 - 6 - 7 658 88
BC014270.2 - 6 - 7 711 88
BT007082.1 - 6 - 7 550 88
DC312466.1 - 3 - 4 506 88
L14283.1 - 6 - 7 550 88
Z15108.1 - 6 - 7 532 88

View Full Product Details