The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_182922.2 | NP_891552.1 | 11 - 12 | 1668 | 97 |
XM_005256013.2 | XP_005256070.1 | 10 - 11 | 1483 | 97 | |
XM_011523188.2 | XP_011521490.1 | 11 - 12 | 1558 | 97 | |
XM_017023385.1 | XP_016878874.1 | 9 - 10 | 1621 | 97 | |
XM_017023386.1 | XP_016878875.1 | 9 - 10 | 1426 | 97 | |
GenBank mRNA | AF462442.1 | - | 8 - 9 | 1860 | 97 |
AK000725.1 | - | 10 - 11 | 1475 | 97 | |
AK289949.1 | - | 11 - 12 | 1633 | 97 | |
BC014415.1 | - | - | 1645 | 97 | |
BC018730.1 | - | 11 - 12 | 1633 | 97 | |
BC033077.1 | - | 11 - 12 | 1534 | 97 |