Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ERBB3
Entrez Gene ID
2065
Gene Name
erb-b2 receptor tyrosine kinase 3
Gene Aliases
ErbB-3, HER3, LCCS2, MDA-BF-1, c-erbB-3, c-erbB3, erbB3-S, p180-ErbB3, p45-sErbB3, p85-sErbB3
Chromosome Location
Chr.12: 56080025 - 56103507 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001982.3 NP_001973.2 26 - 27 3478 66
GenBank mRNA AK124710.1 - 9 - 10 1660 66
AK125028.1 - 4 - 5 1020 66
AK291681.1 - 26 - 27 3331 66
AK294719.1 - 25 - 26 3518 66
AK295650.1 - 27 - 28 3245 66
AK300909.1 - 13 - 14 1577 66
BC002706.2 - 9 - 10 1149 66
BC041579.1 - 11 - 12 1123 66
BC082992.1 - 26 - 27 3317 66
M29366.1 - 26 - 27 3301 66
M34309.1 - 26 - 27 66

View Full Product Details