The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013358.2 | NP_037490.2 | 12 - 13 | 1549 | 110 |
XM_017001101.1 | XP_016856590.1 | 12 - 13 | 1452 | 110 | |
XM_017001102.1 | XP_016856591.1 | 12 - 13 | 1452 | 110 | |
XM_017001103.1 | XP_016856592.1 | 12 - 13 | 1552 | 110 | |
GenBank mRNA | AK026652.1 | - | 12 - 13 | 1551 | 110 |
AK225541.1 | - | 12 - 13 | 1551 | 110 | |
AK293275.1 | - | 2 - 3 | 303 | 110 | |
AK298538.1 | - | 2 - 3 | 300 | 110 | |
AK314644.1 | - | 12 - 13 | 1550 | 110 | |
BC130574.1 | - | 12 - 13 | 1528 | 110 | |
BC136402.1 | - | 12 - 13 | 1528 | 110 |