The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042385.2 | NP_001035844.1 | 7 - 8 | 1195 | 75 |
NM_001171603.1 | NP_001165074.1 | 9 - 10 | 1721 | 75 | |
NM_001171606.1 | NP_001165077.1 | 9 - 10 | 1852 | 75 | |
NM_001171613.1 | NP_001165084.1 | 8 - 9 | 1310 | 75 | |
NM_001171617.1 | NP_001165088.1 | 8 - 9 | 1467 | 75 | |
NM_006036.4 | NP_006027.2 | 8 - 9 | 2306 | 75 | |
XM_011533198.1 | XP_011531500.1 | 9 - 10 | 1808 | 75 | |
XM_011533200.1 | XP_011531502.1 | 9 - 10 | 1460 | 75 | |
XM_011533202.1 | XP_011531504.1 | 8 - 9 | 1260 | 75 | |
XM_017005384.1 | XP_016860873.1 | 9 - 10 | 2446 | 75 | |
XM_017005385.1 | XP_016860874.1 | 9 - 10 | 1771 | 75 | |
GenBank mRNA | AB007896.1 | - | 8 - 9 | 1233 | 75 |
AK025649.1 | - | 8 - 9 | 1153 | 75 | |
AK131463.1 | - | 9 - 10 | 1664 | 75 | |
BC151236.1 | - | 9 - 10 | 1785 | 75 | |
DQ023503.1 | - | 8 - 9 | 2306 | 75 | |
DQ023504.1 | - | 8 - 9 | 2306 | 75 | |
DQ023505.1 | - | 8 - 9 | 2306 | 75 | |
DQ023506.1 | - | 7 - 8 | 1195 | 75 |