The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015431.3 | NP_056246.3 | 1 - 2 | 471 | 152 |
GenBank mRNA | AF327057.1 | - | 1 - 2 | 471 | 152 |
AK096188.1 | - | 1 - 2 | 353 | 152 | |
DN998591.1 | - | 1 - 2 | 470 | 152 |