Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CNR2
Entrez Gene ID
1269
Gene Name
cannabinoid receptor 2
Gene Aliases
CB-2, CB2, CX5
Chromosome Location
Chr.1: 23870526 - 23913327 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA EU517121.1 - 2 - 3 213 86

View Full Product Details