The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286769.1 | NP_001273698.1 | 4 - 5 | 335 | 77 |
NM_030580.4 | NP_085057.3 | 4 - 5 | 331 | 77 | |
XM_011517314.2 | XP_011515616.1 | 4 - 5 | 1658 | 77 | |
XM_011517315.2 | XP_011515617.1 | 4 - 5 | 575 | 77 | |
XM_011517316.2 | XP_011515618.1 | 5 - 6 | 380 | 77 | |
XM_011517318.2 | XP_011515620.1 | 4 - 5 | 1658 | 77 | |
XM_017013872.1 | XP_016869361.1 | 5 - 6 | 381 | 77 | |
XM_017013873.1 | XP_016869362.1 | 4 - 5 | 321 | 77 | |
XM_017013874.1 | XP_016869363.1 | 4 - 5 | 355 | 77 | |
XM_017013875.1 | XP_016869364.1 | 5 - 6 | 521 | 77 | |
GenBank mRNA | AL833814.1 | - | 4 - 5 | 315 | 77 |
BC004480.2 | - | 4 - 5 | 290 | 77 |