The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001251987.1 | NP_001238916.1 | 20 - 21 | 70 | |
NM_006073.3 | NP_006064.2 | 20 - 21 | 70 | ||
GenBank mRNA | AJ489257.2 | - | 20 - 21 | 1541 | 70 |
CR749591.1 | - | 10 - 11 | 402 | 70 | |
U18985.1 | - | 20 - 21 | 1347 | 70 |