The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006073.3 | NP_006064.2 | 22 - 23 | 1735 | 115 |
GenBank mRNA | U18985.1 | - | 22 - 23 | 1439 | 115 |