The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006073.3 | NP_006064.2 | 29 - 30 | 1986 | 145 |
GenBank mRNA | CR749591.1 | - | 18 - 19 | 694 | 145 |
U18985.1 | - | 29 - 30 | 1690 | 145 |